CAS RN 349-76-8
IUPAC Name:
1-[3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms:
m-(trifluoromethyl)benzoyl
Molecular Weight (g/mol):
188.15
Molecular Formula:
C9H7F3O
InChi Key:
ABXGMGUHGLQMAW-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3'-(Trifluoromethyl)acetophenone, 98+%
CAS: 349-76-8 Molecular Formula: C9H7F3O Molecular Weight (g/mol): 188.149 MDL Number: MFCD00000391 InChI Key: ABXGMGUHGLQMAW-UHFFFAOYSA-N Synonym: 3'-trifluoromethyl acetophenone,3-trifluoromethyl acetophenone,3-trifluoromethylacetophenone,ethanone, 1-3-trifluoromethyl phenyl,1-3-trifluoromethyl phenyl ethanone,m-trifluoromethylacetophenone,3'-trifluoromethylacetophenone,1-3-trifluoromethyl phenyl ethan-1-one,1-3-trifluoromethyl-phenyl-ethanone PubChem CID: 67682 IUPAC Name: 1-[3-(trifluoromethyl)phenyl]ethanone SMILES: CC(=O)C1=CC(=CC=C1)C(F)(F)F