accessibility menu, dialog, popup

UserName

SOONH2FFFBr

CAS RN 351003-64-0

IUPAC Name: 4-bromo-3-(trifluoromethyl)benzene-1-sulfonamide
Synonyms: 4-bromo-3-(trifluoromethyl)benzenesulfonamide
Molecular Weight (g/mol): 304.08
Molecular Formula: C7H5BrF3NO2S
InChi Key: GQUUOLHRLZCAIO-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(=C(Br)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results