accessibility menu, dialog, popup

UserName

BNSiH3CCH3H3CH3CCH3OHHO

CAS RN 351457-64-2

IUPAC Name: [1-(tert-butyldimethylsilyl)-1H-indol-4-yl]boronic acid
Synonyms: 1-(tert-butyldimethylsilyl)indol-4-ylboronic acid
Molecular Weight (g/mol): 275.23
Molecular Formula: C14H22BNO2Si
InChi Key: PTZKCFPZTHUQTI-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)N1C=CC2=C(C=CC=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results