missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)indole-4-boronic acid, 98%, Thermo Scientific™

Product Code. 15490316
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490316 1 g 1g
15400326 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490316 Supplier Thermo Scientific Alfa Aesar Supplier No. H53035.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351457-64-2
Molecular Formula C14H22BNO2Si
Molecular Weight (g/mol) 275.23
MDL Number MFCD08436007
InChI Key PTZKCFPZTHUQTI-UHFFFAOYSA-N
Synonym 1-tert-butyldimethylsilyl-1h-indole-4-boronic acid, 1-tert-butyldimethylsilyl indol-4-ylboronic acid, 1-tert-butyldimethylsilyl-1h-indol-4-yl boronic acid, 1-tert-butyldimethylsilyl-1h-indol-4-ylboronic acid, 1-tbdms-indole-4-boronic acid, 1-tert-butyldimethylsilyl indole-4-boronic acid, 1-tert-butyl dimethylsilyl-1h-indole-4-boronic acid, 1-tert-butyl dimethyl silyl-1h-indol-4-yl boronic acid, 1-tert-butyldimethylsilyl-1h-indole-4-boronic acid 98, 1-1,1-dimethylethyl dimethyl silyl-1h-indol-4-yl boronic acid
PubChem CID 22352397
IUPAC Name [1-[tert-butyl(dimethyl)silyl]indol-4-yl]boronic acid
SMILES B(C1=C2C=CN(C2=CC=C1)[Si](C)(C)C(C)(C)C)(O)O

Specifications

Melting Point 130°C to 135°C
Quantity 1 g
Formula Weight 275.23
Percent Purity 98%
Chemical Name or Material 1-(tert-Butyldimethylsilyl)indole-4-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.