accessibility menu, dialog, popup

UserName

CAS RN 358-67-8

CH3OSiCH3FFFOH3C

CAS RN 358-67-8

IUPAC Name: dimethoxy(methyl)(3,3,3-trifluoropropyl)silane
Synonyms: dimethoxy(methyl)(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol): 202.25
Molecular Formula: C6H13F3O2Si
InChi Key: DIJRHOZMLZRNLM-UHFFFAOYSA-N
SMILES: CO[Si](C)(CCC(F)(F)F)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results