CAS RN 35975-00-9
IUPAC Name:
6-nitroquinolin-5-amine
Synonyms:
5-quinolinamine, 6-nitro-
Molecular Weight (g/mol):
189.17
Molecular Formula:
C9H7N3O2
InChi Key:
TYBYHEXFKFLRFT-UHFFFAOYSA-N
SMILES:
NC1=C(C=CC2=NC=CC=C12)[N+]([O-])=O
1
–
3
of
3
results
1
5-Amino-6-nitroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Amino-6-nitroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Amino-6-nitroquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.