CAS RN 359860-27-8
CAS RN 359860-27-8
IUPAC Name:
N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms:
N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol):
418.55
Molecular Formula:
C18H34N4O5S
InChi Key:
KIJSBKNJFAUJFV-UHFFFAOYNA-N
SMILES:
NCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
1
–
4
of
4
results
1
(+)-Biotinyl 3,6,9-Trioxaundecanediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-Biotinyl 3,6,9-Trioxaundecanediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+)-Biotinyl 3,6,9-Trioxaundecanediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
MedChemExpress Amine-PEG3-Biotin
MedChemExpress Amine-PEG3-Biotin is a signal amplification label containing a biotin group and a terminal primary amine group.