accessibility menu, dialog, popup

UserName

CAS RN 36596-67-5

H3COOH3CCH3

CAS RN 36596-67-5

IUPAC Name: ethyl 2,6-dimethylbenzoate
Synonyms: ethyl 2,6-dimethylbenzoate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: NBBSIMLSNFENNF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=CC=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)