Learn More
Ethyl 2,6-dimethylbenzoate, 98%, Thermo Scientific™
Product Code. 11331887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331887 5 g 5g
11321887 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11331887 Supplier Thermo Scientific Alfa Aesar Supplier No. L07381.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36596-67-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00051785
InChI Key NBBSIMLSNFENNF-UHFFFAOYSA-N
Synonym acmc-1ajem, 2,6-dimethylbenzoic acid ethyl ester, 2,6-dimethyl benzoic acid ethyl ester, benzoic acid, 2,6-dimethyl-, ethyl ester
PubChem CID 520807
IUPAC Name ethyl 2,6-dimethylbenzoate
SMILES CCOC(=O)C1=C(C=CC=C1C)C
Specifications
Quantity 5 g
Refractive Index 1.498
Beilstein 2575407
Boiling Point 78°C to 80°C (0.9 mmHg)
Solubility Information Difficult to mix.
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material Ethyl 2,6-dimethylbenzoate