CAS RN 36764-94-0
CAS RN 36764-94-0
IUPAC Name:
2-amino-3,5-dichlorobenzonitrile
Synonyms:
2-amino-3,5-dichlorobenzonitrile
Molecular Weight (g/mol):
187.02
Molecular Formula:
C7H4Cl2N2
InChi Key:
ZHKNDJRPOVUPMT-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=C(Cl)C=C1C#N
1
–
1
of
1
results
1
2-Amino-3,5-dichlorobenzonitrile, 97%, Thermo Scientific™
CAS: 36764-94-0 Molecular Formula: C7H4Cl2N2 Molecular Weight (g/mol): 187.02 MDL Number: MFCD00158947 InChI Key: ZHKNDJRPOVUPMT-UHFFFAOYSA-N Synonym: 2-amino-3,5-dichloro-benzonitrile,2-amino-3,5-dichlorobenzenecarbonitrile,pubchem11850,3,5-dichloroanthranilonitrile,buttpark 52\14-89,benzonitrile,2-amino-3,5-dichloro,2-amino-3,5-dichlorobenzonitrile PubChem CID: 737215 IUPAC Name: 2-amino-3,5-dichlorobenzonitrile SMILES: NC1=C(Cl)C=C(Cl)C=C1C#N