Learn More
2-Amino-3,5-dichlorobenzonitrile, 97%, Thermo Scientific™
Product Code. 15431799
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15431799
Brand: Thermo Scientific Alfa Aesar H64534.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36764-94-0
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.02
MDL Number MFCD00158947
InChI Key ZHKNDJRPOVUPMT-UHFFFAOYSA-N
Synonym 2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile
PubChem CID 737215
IUPAC Name 2-amino-3,5-dichlorobenzonitrile
SMILES NC1=C(Cl)C=C(Cl)C=C1C#N
Specifications
Melting Point 123°C to 127°C (literature)
Quantity 5 g
Beilstein 2720575
Formula Weight 187.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3,5-dichlorobenzonitrile