accessibility menu, dialog, popup

UserName

CAS RN 37486-69-4

FOFFFOFFFOFFFOFFFOFFFFFFFFFFFFFFFFFFFFFF

CAS RN 37486-69-4

IUPAC Name: 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaoctadecane
Synonyms: 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaoctadecane
Molecular Weight (g/mol): 950.13
Molecular Formula: C17HF35O5
InChi Key: MEXQRXXROOSHGK-UHFFFAOYNA-N
SMILES: FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results