Learn More
2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar L16952.14
| Quantity | 25g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 37486-69-4 | |
| 221°C to 225°C | |
| C17HF35O5 | |
| 25g | |
| MEXQRXXROOSHGK-UHFFFAOYNA-N | |
| 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaoctadecane | |
| 123459 | |
| 2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane |
| 1.765 | |
| None | |
| MFCD00054716 | |
| 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 2h-perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane, 3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane, unii-sef36f005e, freon e-5, 2h-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,2,3,3,3-heptakis fluoranyl propoxy propoxy propoxy propoxy-3-1,2,2,2-tetrakis fluoranyl ethoxy propane | |
| FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F | |
| 950.13 | |
| 950.14 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.