accessibility menu, dialog, popup

UserName

BOOCH3H3CCH3CH3HN

CAS RN 388116-27-6

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Molecular Weight (g/mol): 243.11
Molecular Formula: C14H18BNO2
InChi Key: QDCIXBBEUHMLDN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results