accessibility menu, dialog, popup

UserName

CAS RN 3883-94-1

ClH3COONH2

CAS RN 3883-94-1

IUPAC Name: 2-amino-1-(4-methoxyphenyl)ethan-1-one hydrochloridyl
Synonyms: 2-amino-1-(4-methoxyphenyl)ethanone hydrochloridyl
Molecular Weight (g/mol): 200.64
Molecular Formula: C9H11ClNO2
InChi Key: ZSUGWZZUIKUPMO-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=C(C=C1)C(=O)CN
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results