accessibility menu, dialog, popup

UserName

HClClNNH

CAS RN 38869-46-4

IUPAC Name: hydrogen 1-(4-chlorophenyl)piperazine chloride
Synonyms: hydrogen para-chlorophenylpiperazine chloride
Molecular Weight (g/mol): 233.14
Molecular Formula: C10H14Cl2N2
InChi Key: ZHGRQBSZTVJDHU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=C(C=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results