CAS RN 39267-74-8
CAS RN 39267-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Folic Acid Hydrate EP Impurity B (as Sulfate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2,5,6-Triaminopyrimidin-4(1H)-one Sulphate, TRC
CAS: 39267-74-8 Molecular Formula: C4 H7 N5 O . H2 O4 S Molecular Weight (g/mol): 239.21 Synonym: 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate,Folic Acid Imp. B (EP) as Sulphate,Folic Acid Hydrate Impurity B as Sulfate,Folic Acid Impurity B as Sulfate IUPAC Name: sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one SMILES: NC1=NC(=O)C(=C(N)N1)N.OS(=O)(=O)O