CAS RN 3938-96-3
IUPAC Name:
ethyl 2-methoxyacetate
Synonyms:
ethyl 2-methoxyacetate
Molecular Weight (g/mol):
118.13
Molecular Formula:
C5H10O3
InChi Key:
JLEKJZUYWFJPMB-UHFFFAOYSA-N
SMILES:
CCOC(=O)COC
1
–
1
of
1
results
1
Ethyl methoxyacetate, 98%
CAS: 3938-96-3 Molecular Formula: C5H10O3 Molecular Weight (g/mol): 118.132 MDL Number: MFCD00009176 InChI Key: JLEKJZUYWFJPMB-UHFFFAOYSA-N Synonym: ethyl methoxyacetate,acetic acid, methoxy-, ethyl ester,2-methoxy ethyl acetate,ethyl-3-oxabutanoate,methoxyacetic acid ethyl ester,methoxy acetic acid ethyl ester,ethyl-metoxyacetate,methylglycol acetate,ethyl methyloxy acetate,acmc-1cofg PubChem CID: 77544 IUPAC Name: ethyl 2-methoxyacetate SMILES: CCOC(=O)COC