accessibility menu, dialog, popup

UserName

CAS RN 3939-15-9

NFN

CAS RN 3939-15-9

IUPAC Name: 6-fluoropyridine-2-carbonitrile
Synonyms: 6-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: NVOLTPVZQXTZCW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=N1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results