Learn More
2-Cyano-6-fluoropyridine, 95%
Product Code. 15483727
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15483727
Brand: Thermo Scientific Alfa Aesar H27600.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 3939-15-9 | C6H3FN2 | 122.10 g/mol
It is used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as primary and secondary intermediate.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3939-15-9
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.10
MDL Number MFCD09263323
InChI Key NVOLTPVZQXTZCW-UHFFFAOYSA-N
Synonym 2-cyano-6-fluoropyridine, 6-fluoropicolinonitrile, 2-pyridinecarbonitrile, 6-fluoro, 6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile, pubchem4017, acmc-1cj0q, 2-pyridinecarbonitrile,6-fluoro, 6-fluoro-pyridine-2-carbonitrile, 2-fluoropyridine-6-carbonitrile, 6-fluoro-2-pyridinecarbonitrile
PubChem CID 14296369
IUPAC Name 6-fluoropyridine-2-carbonitrile
SMILES FC1=CC=CC(=N1)C#N
Specifications
Melting Point 32°C to 35°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 122.1
Percent Purity 95%
Chemical Name or Material 2-Cyano-6-fluoropyridine