accessibility menu, dialog, popup

UserName

CAS RN 3946-32-5

IUPAC Name: 8-methoxy-8-oxooctanoate
Synonyms: 8-methoxy-8-oxooctanoate
Molecular Weight (g/mol): 187.22
Molecular Formula: C9H15O4
InChi Key: KOVPXZDUVJGGFU-UHFFFAOYSA-M
SMILES: COC(=O)CCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)