CAS RN 39815-78-6
IUPAC Name:
methyl 3-oxoheptanoate
Synonyms:
methyl 3-oxoheptanoate
Molecular Weight (g/mol):
158.2
Molecular Formula:
C8H14O3
InChi Key:
CZTKGERSDUGZPQ-UHFFFAOYSA-N
SMILES:
CCCCC(=O)CC(=O)OC
1
–
3
of
3
results
1
Methyl 3-Oxoheptanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 3-Oxoheptanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 3-Oxoheptanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.