accessibility menu, dialog, popup

UserName

CH3FFF

CAS RN 401-79-6

IUPAC Name: 1-methyl-3-(trifluoromethyl)benzene
Synonyms: 1-methyl-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 160.14
Molecular Formula: C8H7F3
InChi Key: VJYXZJGDFJJDGF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results