accessibility menu, dialog, popup

UserName

CAS RN 40493-18-3

ClNNS

CAS RN 40493-18-3

IUPAC Name: 3-chloro-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Synonyms: 3-chloro-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Molecular Weight (g/mol): 224.71
Molecular Formula: C10H9ClN2S
InChi Key: PRNJDUCSVXTOTN-UHFFFAOYSA-N
SMILES: ClC1=C2C3=C(CCCC3)SC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results