accessibility menu, dialog, popup

UserName

CAS RN 4097-88-5

CH3NH2NNH2

CAS RN 4097-88-5

IUPAC Name: bis(2-aminoethyl)(methyl)amine
Synonyms: bis(2-aminoethyl)(methyl)amine
Molecular Weight (g/mol): 117.2
Molecular Formula: C5H15N3
InChi Key: HYSQEYLBJYFNMH-UHFFFAOYSA-N
SMILES: CN(CCN)CCN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)