missing translation for 'onlineSavingsMsg'
Learn More

N1-(2-Aminoethyl)-N1-Methylethane-1,2-Diamine, TRC

Product Code. 30363876
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363876 250 mg 250mg
30361046 500 mg -
30352716 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363876 Supplier Toronto Research Chemicals Supplier No. TRCD100548250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-88-5
Molecular Formula C5H15N3
Molecular Weight (g/mol) 117.19
Synonym 1,2-Ethanediamine, N-(2-aminoethyl)-N-methyl- (9CI), Diethylenetriamine, 4-methyl- (6CI,7CI,8CI), 1,5-Diamino-3-methyl-3-azapentane, 1,5-Diamino-N'-methyl-3-azapentane, 2,2'-Diamino-N-methyldiethylamine, 3-Methyl-3-Aza-pentane-1,5-diamine, 4-Methyl-1,4,7-triazaheptane, 4-Methyldiethylenetriamine, Bis(2-aminoethyl)methylamine, Di(2-aminoethyl)methylamine, N,N-Bis(2-aminoethyl)methylamine, N,N-Bis(beta-aminoethyl)methylamine, N-(2-Aminoethyl)-N-methyl-1,2-ethanediamine, N-(2-Aminoethyl)-N-methylethylenediamine, N-Methyl-2,2'-diaminodiethylamine, N1-(2-Aminoethyl)-N1-methylethane-1,2-diamine, N1-(2-Aminoethyl)-N1-methylethane-1,2-diamine, N2-Methyldiethylenetriamine, N'-Methyl-2,2'-diaminodiethylamine, N'-Methyldiethylenetriamine
IUPAC Name N'-(2-aminoethyl)-N'-methylethane-1,2-diamine
SMILES CN(CCN)CCN

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H15N3/c1-8(4-2-6)5-3-7/h2-7H2,1H3
Formula Weight 117.1266
Chemical Name or Material N1-(2-Aminoethyl)-N1-Methylethane-1,2-Diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.