accessibility menu, dialog, popup

UserName

CAS RN 4265-16-1

OO

CAS RN 4265-16-1

IUPAC Name: 1-benzofuran-2-carbaldehyde
Synonyms: 1-benzofuran-2-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: ADDZHRRCUWNSCS-UHFFFAOYSA-N
SMILES: O=CC1=CC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)