accessibility menu, dialog, popup

UserName

ClOClCl

CAS RN 42981-08-8

IUPAC Name: 2-chloro-1-(3,4-dichlorophenyl)ethan-1-one
Synonyms: 2-chloro-1-(3,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 223.48
Molecular Formula: C8H5Cl3O
InChi Key: BYTZWANJVUAPNO-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)