accessibility menu, dialog, popup

UserName

H3COBrO

CAS RN 43192-33-2

IUPAC Name: 4-bromo-2-methoxybenzaldehyde
Synonyms: 4-bromo-2-methoxybenzaldehyde
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: YMXQYZABFWVXEK-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results