missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 15491838
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491838 1 g 1g
15411848 250 mg 250mg
15401848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491838 Supplier Thermo Scientific Alfa Aesar Supplier No. H55545.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromo-2-methoxybenzaldehyde is used in the building block, synthesis of a nonacid degradable linker for solid-phase synthesis.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43192-33-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD08276823
InChI Key YMXQYZABFWVXEK-UHFFFAOYSA-N
PubChem CID 11053041
IUPAC Name 4-bromo-2-methoxybenzaldehyde
SMILES COC1=C(C=O)C=CC(Br)=C1

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Sensitivity Air Sensitive
Synonym 4-bromo-2-methoxy-benzaldehyde, benzaldehyde, 4-bromo-2-methoxy, 2-methoxy-4-bromobenzaldehyde, 4-bromo-2-methoxy benzaldehyde, acmc-209jte, 4-bromo-2-methoxybezaldehyde, ksc494s3l, 4-bromo-2-methoxybenz-aldehyde, benzaldehyde,4-bromo-2-methoxy, 4-bromanyl-2-methoxy-benzaldehyde
Solubility Information Soluble in water.
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.