accessibility menu, dialog, popup

UserName

CAS RN 4766-57-8

H3COSiOCH3OCH3OCH3

CAS RN 4766-57-8

IUPAC Name: tetrabutyl silicate
Synonyms: tetrabutyl silicate
Molecular Weight (g/mol): 320.55
Molecular Formula: C16H36O4Si
InChi Key: UQMOLLPKNHFRAC-UHFFFAOYSA-N
SMILES: CCCCO[Si](OCCCC)(OCCCC)OCCCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results