accessibility menu, dialog, popup

UserName

CAS RN 4792-15-8

HOOOOOOH

CAS RN 4792-15-8

IUPAC Name: 3,6,9,12-tetraoxatetradecane-1,14-diol
Synonyms: pentaethylene glycol
Molecular Weight (g/mol): 238.28
Molecular Formula: C10H22O6
InChi Key: JLFNLZLINWHATN-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCO
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)