accessibility menu, dialog, popup

UserName

CAS RN 491-37-2

OO

CAS RN 491-37-2

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: MSTDXOZUKAQDRL-UHFFFAOYSA-N
SMILES: O=C1CCOC2=CC=CC=C12
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)