CAS RN 49669-13-8
CAS RN 49669-13-8
IUPAC Name:
1-(6-bromopyridin-2-yl)ethan-1-one
Synonyms:
1-(6-bromopyridin-2-yl)ethanone
Molecular Weight (g/mol):
200.04
Molecular Formula:
C7H6BrNO
InChi Key:
RUJTWTUYVOEEFW-UHFFFAOYSA-N
SMILES:
CC(=O)C1=NC(Br)=CC=C1
1
–
1
of
1
results
1
2-Acetyl-6-bromopyridine, 97%
CAS: 49669-13-8 Molecular Formula: C7H6BrNO Molecular Weight (g/mol): 200.035 MDL Number: MFCD00272200 InChI Key: RUJTWTUYVOEEFW-UHFFFAOYSA-N Synonym: 2-acetyl-6-bromopyridine,2-bromo-6-acetylpyridine,1-6-bromopyridin-2-yl ethanone,1-6-bromopyridin-2-yl ethan-1-one,1-6-bromo-2-pyridinyl-1-ethanone,ethanone, 1-6-bromo-2-pyridinyl,bromopyridinylethanone,pubchem13351,acmc-1ap21,ksc490s1f PubChem CID: 11298578 IUPAC Name: 1-(6-bromopyridin-2-yl)ethanone SMILES: CC(=O)C1=NC(=CC=C1)Br