accessibility menu, dialog, popup

UserName

H3COHOCH3O

CAS RN 498-02-2

IUPAC Name: 1-(4-hydroxy-3-methoxyphenyl)ethan-1-one
Synonyms: apocynin acetovanilone acetovanillone acetoguaiacone apocynine 3-metoksy-4-hydroksyacetofenon acetovanyllon acetoguaiacon
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DFYRUELUNQRZTB-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1O)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)