CAS RN 50-31-7
IUPAC Name:
2,3,6-trichlorobenzoic acid
Synonyms:
2,3,6-trichlorobenzoic acid
Molecular Weight (g/mol):
225.45
Molecular Formula:
C7H3Cl3O2
InChi Key:
XZIDTOHMJBOSOX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Cl)C=CC(Cl)=C1Cl
1
–
3
of
3
results
1
2,3,6-Trichlorobenzoic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3,6-Trichlorobenzoic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3,6-Trichlorobenzoic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.