accessibility menu, dialog, popup

UserName

CAS RN 502-65-8

CH3CH3CH3CH3CH3H3CH3CH3CH3CH3C(E)(E)(E)(E)(E)(E)(E)(E)(E)(E)(E)

CAS RN 502-65-8

IUPAC Name: (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
Synonyms: lycopene lycopene, (cis)-isomer lyc-o-mato (cis)-lycopene (cis)-isomer lycopene lyc-O-mato lycopene, all-trans- (8CI) (13-cis)-lycopene lyco vit lycopene 7
Molecular Weight (g/mol): 536.89
Molecular Formula: C40H56
InChi Key: OAIJSZIZWZSQBC-GYZMGTAESA-N
SMILES: CC(C)=CCC\C(C)=C\C=C\C(\C)=C\C=C\C(\C)=C\C=C\C=C(/C)\C=C\C=C(/C)\C=C\C=C(/C)CCC=C(C)C
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)