accessibility menu, dialog, popup

UserName

CAS RN 50342-01-3

NOCl

CAS RN 50342-01-3

IUPAC Name: quinoline-2-carbonyl chloride
Synonyms: quinoline-2-carbonyl chloride
Molecular Weight (g/mol): 191.61
Molecular Formula: C10H6ClNO
InChi Key: WFVMVMAUXYOQSW-UHFFFAOYSA-N
SMILES: ClC(=O)C1=NC2=CC=CC=C2C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results