CAS RN 50390-76-6
CAS RN 50390-76-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Methoxy-5-(methylsulfonyl)benzoic Acid, TRC
CAS: 50390-76-6 Molecular Formula: C9 H10 O5 S Molecular Weight (g/mol): 230.24 Synonym: 2-Methoxy-5-(methylsulfonyl)benzoic acid,5-Methylsulfonyl-2-methoxybenzoic acid,6-Methoxy-3-methylsulfonylbenzoic acid,5-mesyl-o-anisic acid,Tiapride Hydrochloride Impurity B (EP) IUPAC Name: 2-methoxy-5-methylsulfonylbenzoic acid SMILES: COc1ccc(cc1C(=O)O)S(=O)(=O)C
2
Tiapride EP Impurity B, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.