accessibility menu, dialog, popup

UserName

CAS RN 504-01-8

OHHO

CAS RN 504-01-8

IUPAC Name: cyclohexane-1,3-diol
Synonyms: cyclohexane-1,3-diol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: RLMGYIOTPQVQJR-UHFFFAOYNA-N
SMILES: OC1CCCC(O)C1
Boiling Point 
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)