CAS RN 50634-05-4
CAS RN 50634-05-4
IUPAC Name:
2,5-dimethoxyoxolane-3-carbaldehyde
Synonyms:
2,5-dimethoxyoxolane-3-carbaldehyde
Molecular Weight (g/mol):
160.17
Molecular Formula:
C7H12O4
InChi Key:
QMIGEDXMDGEZSR-UHFFFAOYNA-N
SMILES:
COC1CC(C=O)C(OC)O1
1
–
3
of
3
results
1
2,5-Dimethoxytetrahydro-3-furancarboxaldehyde, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,5-Dimethoxytetrahydro-3-furancarboxaldehyde, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,5-Dimethoxytetrahydro-3-furancarboxaldehyde, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.