accessibility menu, dialog, popup

UserName

CAS RN 50800-92-5

H3COONH3

CAS RN 50800-92-5

IUPAC Name: 2-(4-methoxyphenoxy)ethan-1-aminium
Synonyms: 2-(4-methoxyphenoxy)ethanaminium
Molecular Weight (g/mol): 168.22
Molecular Formula: C9H14NO2
InChi Key: YXRCDWIZAGWUBL-UHFFFAOYSA-O
SMILES: COC1=CC=C(OCC[NH3+])C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)