CAS RN 5197-87-5
IUPAC Name:
benzyltripropylazanium chloride
Synonyms:
benzyltripropylazanium chloride
Molecular Weight (g/mol):
269.86
Molecular Formula:
C16H28ClN
InChi Key:
YTRIOKYQEVFKGU-UHFFFAOYSA-M
SMILES:
[Cl-].CCC[N+](CCC)(CCC)CC1=CC=CC=C1