CAS RN 52119-39-8
IUPAC Name:
ethyl 3-(2-nitrophenyl)-3-oxopropanoate
Synonyms:
ethyl 3-(2-nitrophenyl)-3-oxopropanoate
Molecular Weight (g/mol):
237.21
Molecular Formula:
C11H11NO5
InChi Key:
OWZNCVIBJQPNEF-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)C1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Ethyl 2-nitrobenzoylacetate, 97%
CAS: 52119-39-8 Molecular Formula: C11H11NO5 Molecular Weight (g/mol): 237.21 MDL Number: MFCD03424821 InChI Key: OWZNCVIBJQPNEF-UHFFFAOYSA-N Synonym: ethyl 3-2-nitrophenyl-3-oxopropanoate,ethyl 2-nitrobenzoylacetate,acmc-1awhb,ethyl 3-2-nitrophenyl-3-oxo-propanoate,ethyl o-nitrobenzoylacetate,ethyl 2-o-nitrobenzoyl acetate,ethyl 2-2-nitrobenzoyl acetate,ethyl 2-o-nitrobenzoyl-acetate,ethyl 2-nitrobenzoyl acetate PubChem CID: 10561837 IUPAC Name: ethyl 3-(2-nitrophenyl)-3-oxopropanoate SMILES: CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-]