CAS RN 52669-92-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[4-(4-Chlorophenyl)-3,6-dihydro-1(2H)-pyridinyl]-1-(4-fluorophenyl)-1-butanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.