CAS RN 5273-86-9
CAS RN 5273-86-9
IUPAC Name:
1,2,4-trimethoxy-5-[(1Z)-prop-1-en-1-yl]benzene
Synonyms:
β-asarone cis-asarone cis-isoasarone trans-asarone cis-isoelemicin (Z)-asarone
Molecular Weight (g/mol):
208.26
Molecular Formula:
C12H16O3
InChi Key:
RKFAZBXYICVSKP-WAYWQWQTSA-N
SMILES:
COC1=CC(OC)=C(\C=C/C)C=C1OC
1
–
1
of
1
results
1
cis-beta-Asarone (>70%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.