CAS RN 52807-12-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4’-Hydroxy Flurbiprofen, TRC
CAS: 52807-12-2 Molecular Formula: C15 H13 F O3 Molecular Weight (g/mol): 260.26 Synonym: 4'-Hydroxy Flurbiprofen IUPAC Name: 2-[3-fluoro-4-(4-hydroxyphenyl)phenyl]propanoic acid SMILES: CC(C(=O)O)c1ccc(c(F)c1)c2ccc(O)cc2