missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Flurbiprofen, TRC

Product Code. 30133616
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133616 50 mg 50mg
30008693 5 mg 5mg
30003868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133616 Supplier Toronto Research Chemicals Supplier No. TRCH94244050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52807-12-2
Molecular Formula C15 H13 F O3
Molecular Weight (g/mol) 260.26
Synonym 4'-Hydroxy Flurbiprofen
IUPAC Name 2-[3-fluoro-4-(4-hydroxyphenyl)phenyl]propanoic acid
SMILES CC(C(=O)O)c1ccc(c(F)c1)c2ccc(O)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H13FO3/c1-9(15(18)19)11-4-7-13(14(16)8-11)10-2-5-12(17)6-3-10/h2-9,17H,1H3,(H,18,19)
Formula Weight 260.0849
Percent Purity >95%
Chemical Name or Material 4'-Hydroxy Flurbiprofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.