CAS RN 5368-28-5
IUPAC Name:
3-phenylpiperazin-2-one
Synonyms:
3-phenylpiperazin-2-one
Molecular Weight (g/mol):
176.22
Molecular Formula:
C10H12N2O
InChi Key:
WKFFHKBGGZHQAX-UHFFFAOYNA-N
SMILES:
O=C1NCCNC1C1=CC=CC=C1
1
–
1
of
1
results
1
3-Phenyl-2-piperazinone, 97%, Thermo Scientific™
CAS: 5368-28-5 Molecular Formula: C10H12N2O Molecular Weight (g/mol): 176.22 MDL Number: MFCD01871361 InChI Key: WKFFHKBGGZHQAX-UHFFFAOYNA-N Synonym: 3-phenyl-2-piperazinone,3-phenyl-piperazin-2-one,2-oxo-3-phenylpiperazine,3-phenyl-2-piperazinene,3-phenyl-2-oxopiperazine,3-phenylpiperazine-2-one,2-piperazinone,3-phenyl,2-oxo-3-phenyl-piperazine,3-phenyl-piperazine-2-one,2-phenyl-3-keto piperazine PubChem CID: 250681 IUPAC Name: 3-phenylpiperazin-2-one SMILES: O=C1NCCNC1C1=CC=CC=C1