accessibility menu, dialog, popup

UserName

CAS RN 537013-52-8

FBrFCl

CAS RN 537013-52-8

IUPAC Name: 2-(bromomethyl)-5-chloro-1,3-difluorobenzene
Synonyms: 2-(bromomethyl)-5-chloro-1,3-difluorobenzene
Molecular Weight (g/mol): 241.46
Molecular Formula: C7H4BrClF2
InChi Key: KYKUEAHGMDKVLF-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(F)=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results